Learn More
Apexbio Technology LLC Vatalanib (PTK787) 2HCl 212141-51-0 10mg
SDP
Catalog No. 5000095886
Encompass_Preferred
Vatalanib (PTK787) 2HCl (CAS 212141-51-0) is a small molecule inhibitor targeting vascular endothelial growth factor (VEGF) receptor tyrosine kinases including VEGFR/KDR (IC50 0 037 M) VEGFR/Flt-1 (IC50 0 077 M) and VEGFR/Flk (IC50 0 27 M) It also inhibits other receptor tyrosine kinases such as PDGFR- (IC50 0 58 M) c-Kit (IC50 0 73 M) and c-Fms (IC50 1 4 M) Through blockade of these pathways Vatalanib suppresses VEGF-mediated signaling and angiogenesis It is utilized in research investigating mechanisms of angiogenesis and has potential applications in studies of solid tumors and other angiogenesis-dependent diseases
Catalog No. 50-000-95886 Supplier Apexbio Technology LLC Supplier No. A177810
Vatalanib (PTK787) 2HCl (CAS 212141-51-0) is a small molecule inhibitor targeting vascular endothelial growth factor (VEGF) receptor tyrosine kinases including VEGFR/KDR (IC50 0 037 M) VEGFR/Flt-1 (IC50 0 077 M) and VEGFR/Flk (IC50 0 27 M) It also inhibits other receptor tyrosine kinases such as PDGFR- (IC50 0 58 M) c-Kit (IC50 0 73 M) and c-Fms (IC50 1 4 M) Through blockade of these pathways Vatalanib suppresses VEGF-mediated signaling and angiogenesis It is utilized in research investigating mechanisms of angiogenesis and has potential applications in studies of solid tumors and other angiogenesis-dependent diseases