Learn More
Medchemexpress LLC Vatinoxan hydrochloride | 130466-38-5 | 99.79% | 454.97 | 25 MG
SDP
Catalog No. 5000385556
Encompass_Preferred
Vatinoxan hydrochloride is a peripheral α2 adrenergic receptor antagonist. It has been shown to increase cardiac index and tissue oxygen delivery without deleterious adverse effects when administered alone. Furthermore, it attenuates or prevents dexmedetomidine's systemic hemodynamic effects in a dose-dependent manner and shortens the sedative effect without altering its quality. This product is intended for research use only.
  • Peripheral α2 adrenergic receptor antagonist.
  • Increases cardiac index and tissue oxygen delivery.
  • Attenuates or prevents dexmedetomidine's systemic hemodynamic effects.
  • Shortens sedative effect without altering its quality.
  • Intended for research use only.
Catalog No. 50-003-85556 Supplier Medchemexpress LLC Supplier No. 25MGHY19057A
Vatinoxan hydrochloride is a peripheral α2 adrenergic receptor antagonist. It has been shown to increase cardiac index and tissue oxygen delivery without deleterious adverse effects when administered alone. Furthermore, it attenuates or prevents dexmedetomidine's systemic hemodynamic effects in a dose-dependent manner and shortens the sedative effect without altering its quality. This product is intended for research use only.
  • Peripheral α2 adrenergic receptor antagonist.
  • Increases cardiac index and tissue oxygen delivery.
  • Attenuates or prevents dexmedetomidine's systemic hemodynamic effects.
  • Shortens sedative effect without altering its quality.
  • Intended for research use only.