Learn More
Medchemexpress LLC Vatinoxan (hydrochloride) | 130466-38-5 | 99.8% | 454.97 | 50 MG
SDP
Catalog No. 5000385862
Encompass_Preferred
Vatinoxan hydrochloride is a peripheral α2 adrenergic receptor antagonist. It is for research use only and may be useful in attenuating reductions in heart rate.
  • Increases cardiac index and tissue oxygen delivery without adverse effects.
  • Attenuates or prevents dexmedetomidine's systemic hemodynamic effects in a dose-dependent manner.
  • Induces the least alterations in cardiovascular function at a 50:1 dose ratio (Vatinoxan:dexmedetomidine).
  • Attenuates bradycardia associated with dexmedetomidine.
  • Shortens sedative effect without altering its quality.
Catalog No. 50-003-85862 Supplier Medchemexpress LLC Supplier No. HY19057A50MG
Vatinoxan hydrochloride is a peripheral α2 adrenergic receptor antagonist. It is for research use only and may be useful in attenuating reductions in heart rate.
  • Increases cardiac index and tissue oxygen delivery without adverse effects.
  • Attenuates or prevents dexmedetomidine's systemic hemodynamic effects in a dose-dependent manner.
  • Induces the least alterations in cardiovascular function at a 50:1 dose ratio (Vatinoxan:dexmedetomidine).
  • Attenuates bradycardia associated with dexmedetomidine.
  • Shortens sedative effect without altering its quality.