Learn More
Cayman Chemical VatlnIbhydrochlorIde 100mg
SDP
Catalog No. 502973188
Encompass_Preferred
An antagonist of VEGFR1, VEGFR2, and VEGFR3 with IC50 values of 77, 37, and 190 nM, respectively; less potently inhibits PDGF and c-Kit (IC50 = 600 and 700 nM) and has no effect on additional kinases; completely blocks retinal neovascularization in oxygen-induced ischemic retinopathy in mice
Catalog No. 50-297-3188 Supplier Cayman Chemical Supplier No. 14868100 MG
An antagonist of VEGFR1, VEGFR2, and VEGFR3 with IC50 values of 77, 37, and 190 nM, respectively; less potently inhibits PDGF and c-Kit (IC50 = 600 and 700 nM) and has no effect on additional kinases; completely blocks retinal neovascularization in oxygen-induced ischemic retinopathy in mice