Learn More
Cayman Chemical Vatalanib (hydrochloride), Purity: ≥98%, CAS: 212141-51-0, 50 mg
SDP
Catalog No. 502971554
Encompass_Preferred
An antagonist of VEGFR1, VEGFR2, and VEGFR3 with IC50 values of 77, 37, and 190 nM, respectively; less potently inhibits PDGF and c-Kit (IC50 = 600 and 700 nM) and has no effect on additional kinases; completely blocks retinal neovascularization in oxygen-induced ischemic retinopathy in mice
Catalog No. 50-297-1554 Supplier Cayman Chemical Supplier No. 1486850 MG
An antagonist of VEGFR1, VEGFR2, and VEGFR3 with IC50 values of 77, 37, and 190 nM, respectively; less potently inhibits PDGF and c-Kit (IC50 = 600 and 700 nM) and has no effect on additional kinases; completely blocks retinal neovascularization in oxygen-induced ischemic retinopathy in mice