Learn More
Cayman Chemical Vatqunon 10mg
SDP
Catalog No. 502935313
Encompass_Preferred
A metabolite of α-tocotrienol that has antioxidant and ferroptosis inhibitory activity; inhibits BSO-induced decreases in cell in primary fibroblasts isolated from patients with Friedreich's ataxia or Leigh syndrome (EC50s = 24 and 30 nM, respectively); inhibits RSL3-induced cell death and lipid peroxidation in primary fibroblasts isolated from patients with PCH6 (EC50s = 17.3-21.8 and 33.1-57.6 nM, respectively)
Catalog No. 50-293-5313 Supplier Cayman Chemical Supplier No. 2681910 MG
A metabolite of α-tocotrienol that has antioxidant and ferroptosis inhibitory activity; inhibits BSO-induced decreases in cell in primary fibroblasts isolated from patients with Friedreich's ataxia or Leigh syndrome (EC50s = 24 and 30 nM, respectively); inhibits RSL3-induced cell death and lipid peroxidation in primary fibroblasts isolated from patients with PCH6 (EC50s = 17.3-21.8 and 33.1-57.6 nM, respectively)