Learn More
Aviva Systems Biology Corp Vax1 Antibody - N-terminal region (ARP37000_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37000_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Vax1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MGC130490ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-56308