Learn More
Selleck Chemical LLC Vazegepant hydrochloride
SDP
Catalog No. 503138962
Encompass_Preferred
Vazegepant hydrochloride (BHV-3500 Zavegepant) is a highly potent selective and structurally unique small molecule antagonist of CGRP receptor It can also be used for the acute treatment of migraine
Catalog No. 50-313-8962 Supplier Selleck Chemical LLC Supplier No. E182425MG
Vazegepant hydrochloride (BHV-3500 Zavegepant) is a highly potent selective and structurally unique small molecule antagonist of CGRP receptor It can also be used for the acute treatment of migraine