Learn More
TARGETMOL CHEMICALS INC Vazegepant hydrochloride 1MG
SDP
Catalog No. 502876424
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vazegepant hydrochloride (Vazegepant HCl) is the first intranasal gepant for the acute treatment of migraine and has announced a positive phase II/III study. Purity 99.56%
Catalog No. 50-287-6424 Supplier TARGETMOL CHEMICALS INC Supplier No. T86821MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vazegepant hydrochloride (Vazegepant HCl) is the first intranasal gepant for the acute treatment of migraine and has announced a positive phase II/III study. Purity 99.56%