Learn More
Selleck Chemical LLC Vazegepant hydrochloride
Supplier: Selleck Chemical LLC E18241G
Vazegepant hydrochloride (BHV-3500 Zavegepant) is a highly potent selective and structurally unique small molecule antagonist of CGRP receptor It can also be used for the acute treatment of migraine