Learn More
DSI LLC GC Column VB-5 column, VB-5 column, 60m x 0.32mm ID x 5.00df
Catalog No. 502537453
Encompass_Preferred
GC Column VB-5 column, VB-5 column, 60m x 0.32mm ID x 5.00df
Catalog No. 50-253-7453 Supplier DSI LLC Supplier No. CFSA06032200B
GC Column VB-5 column, VB-5 column, 60m x 0.32mm ID x 5.00df