Learn More
DSI LLC VB-35 Column, 30m x 0.25mm ID, CFS, C03025-025B
Catalog No. 502537377
Encompass_Preferred
VB-35 Column, 30m x 0.25mm ID, CFS, C03025-025B
Catalog No. 50-253-7377 Supplier DSI LLC Supplier No. CFSC03025025B
VB-35 Column, 30m x 0.25mm ID, CFS, C03025-025B