Learn More
DSI LLC GC Column FS Valcobond VB 5, 15m Length, ID 0.32mm, DF 5.00um, CFS-B01532-500B
Supplier:  DSI LLC CFSB01532500B
Encompass_Preferred
GC Column FS Valcobond VB 5, 15m Length, ID 0.32mm, DF 5.00um, CFS-B01532-500B
Catalog No. 50-253-7430