Learn More
DSI LLC GC Column FS Valcobond VB 5, 15m Length, ID 0.32mm, df 1.00um, CFS-B01532-100B
Supplier:  DSI LLC CFSB01553100B
Encompass_Preferred
GC Column FS Valcobond VB 5, 15m Length, ID 0.32mm, df 1.00um, CFS-B01532-100B
Catalog No. 50-253-7428