Learn More
DSI LLC FS ValcoBond VB-5, 30m Length, ID 0.25mm, DF 0.25um, CFS-B03025-050B
Catalog No. 502537416
Encompass_Preferred
FS ValcoBond VB-5, 30m Length, ID 0.25mm, DF 0.25um, CFS-B03025-050B
Catalog No. 50-253-7416 Supplier DSI LLC Supplier No. CFSB03025050B
FS ValcoBond VB-5, 30m Length, ID 0.25mm, DF 0.25um, CFS-B03025-050B