Learn More
DSI LLC GC Column FS ValcoBond VB 5, 30m Length, ID 0.25mm, DF 0.25um, CFS B03025-025B
Catalog No. 502537417
Encompass_Preferred
GC Column FS ValcoBond VB 5, 30m Length, ID 0.25mm, DF 0.25um, CFS B03025-025B
Catalog No. 50-253-7417 Supplier DSI LLC Supplier No. CFSB03025025B
GC Column FS ValcoBond VB 5, 30m Length, ID 0.25mm, DF 0.25um, CFS B03025-025B