Learn More
DSI LLC GC Column FS Valcobond VB 50, 608, 30m Length, ID 0.53mm, DF 0.50um, CFS-D03053-050B
Catalog No. 502537360
Encompass_Preferred
GC Column FS Valcobond VB 50, 608, 30m Length, ID 0.53mm, DF 0.50um, CFS-D03053-050B
Catalog No. 50-253-7360 Supplier DSI LLC Supplier No. CFSD03025050B
GC Column FS Valcobond VB 50, 608, 30m Length, ID 0.53mm, DF 0.50um, CFS-D03053-050B