Learn More
Selleck Chemical LLC VB124-5mg
SDP
Catalog No. 502389775
Encompass_Preferred
VB124, a potent and selective MCT4 inhibitor, specifically inhibits lactate efflux/import in MDA-MB-231 cells expressing MCT4, and is used for the research of cardiac hypertrophy, heart failure, and metabolism. 
Catalog No. 50-238-9775 Supplier Selleck Chemical LLC Supplier No. S99295MG
VB124, a potent and selective MCT4 inhibitor, specifically inhibits lactate efflux/import in MDA-MB-231 cells expressing MCT4, and is used for the research of cardiac hypertrophy, heart failure, and metabolism. 