Learn More
Biomatik Corporation VB2 / Vitamin B2 ELISA Kit
Supplier:  Biomatik Corporation EKN52635
Encompass_Preferred
Species Reactivity: General species, Format: 96-Well Plate, Assay Type: Competitive Inhibition, Assay Time: 2 hours, UniProt ID: , Restriction: For Research Use Only. Not for use in diagnostic procedures. Not for human or animal drug or food use.
Catalog No. NC2072292