Learn More
Selleck Chemical LLC VBIT-12-5mg
SDP
Catalog No. 502390010
Encompass_Preferred
VBIT-12 is a potent inhibitor of Voltage-dependent anion-selective channel 1 (VDAC1), directly interacting with purified VDAC1, reducing its channel conductance and preventing VDAC1 oligomerization.
Catalog No. 50-239-0010 Supplier Selleck Chemical LLC Supplier No. S89365MG
VBIT-12 is a potent inhibitor of Voltage-dependent anion-selective channel 1 (VDAC1), directly interacting with purified VDAC1, reducing its channel conductance and preventing VDAC1 oligomerization.