Learn More
Selleck Chemical LLC VBIT-4 S3544-100mg
SDP
Catalog No. 503109241
Encompass_Preferred
VBIT-4 is a voltage-dependent anion channel (VDAC) oligomerization inhibitor that decreases mitochondrial DNA (mtDNA) release type I interferon (IFN) signaling neutrophil extracellular traps and disease severity in a mouse model of systemic lupus erythematosus
Catalog No. 50-310-9241 Supplier Selleck Chemical LLC Supplier No. S3544100MG
Add to Cart
Add to Cart
VBIT-4 is a voltage-dependent anion channel (VDAC) oligomerization inhibitor that decreases mitochondrial DNA (mtDNA) release type I interferon (IFN) signaling neutrophil extracellular traps and disease severity in a mouse model of systemic lupus erythematosus