Learn More
Selleck Chemical LLC VBIT-4 5mg
SDP
Supplier:  Selleck Chemical LLC S35445MG
Encompass_Preferred
VBIT-4 is avoltage-dependent anion channel (VDAC) oligomerization inhibitor that decreasesmitochondrial DNA (mtDNA) release,type I interferon (IFN) signaling, neutrophil extracellular traps, and disease severity in a mouse model of systemic lupus erythematosus. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3119