Learn More
CLICKBIO INC VBLOK 200 E-Beamed Irradiated (24/CS)
Catalog No. NC2224824
Encompass
The VBLOK 200 reservoir is a V bottom polypropylene ANSI-SLAS footprint low dead volume reservoir. Designed for automated and manual workflows, it is ideal for nearly complete recovery of samples and reagents. It may also used for recovery of prepared NGS libraries by inverting a PCR plate in the reservior and centrifuging. It is e-beam irradiated for aseptic workflows.
This item is not returnable. View return policy
Catalog No. NC2224824 Supplier CLICKBIO INC Supplier No. VBLOK2001E
The VBLOK 200 reservoir is a V bottom polypropylene ANSI-SLAS footprint low dead volume reservoir. Designed for automated and manual workflows, it is ideal for nearly complete recovery of samples and reagents. It may also used for recovery of prepared NGS libraries by inverting a PCR plate in the reservior and centrifuging. It is e-beam irradiated for aseptic workflows.