Learn More
CLICKBIO INC VBLOK 200 V-Bottom Reservoir (24/CS)
Catalog No. NC1661318
Encompass
The VBLOK 200 reservoir is a V bottom polypropylene ANSI-SLAS footprint low dead volume reservoir. Designed for automated and manual workflows, it is ideal for nearly complete recovery of samples and reagents. It may also used for recovery of prepared NGS libraries by inverting a PCR plate in the reservior and centrifuging.
Catalog No. NC1661318 Supplier CLICKBIO INC Supplier No. VBLOK2001
The VBLOK 200 reservoir is a V bottom polypropylene ANSI-SLAS footprint low dead volume reservoir. Designed for automated and manual workflows, it is ideal for nearly complete recovery of samples and reagents. It may also used for recovery of prepared NGS libraries by inverting a PCR plate in the reservior and centrifuging.