Learn More
Medchemexpress LLC HY-15742 1mg Medchemexpress, Vc-MMAD CAS:1401963-17-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY157421MG
Encompass_Preferred
Medchemexpress, HY-15742 1mg Vc-MMAD CAS:1401963-17-4 Vc-MMAD consists the ADCs linker (Val-Cit) and potent tubulin inhibitor (MMAD). Vc-MMAD is a drug-linker conjugate for ADC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1787