Learn More
MSC MSC VCI Tubing, 8 x 10 in, 4 mil, 1000/Pk.
Catalog No. 501917351
Encompass
This VCI Poly Tubing offers effective protection for items requiring corrosion inhibition during storage. Made from low-density polyethylene film infused with a corrosion inhibitor, it forms a reliable barrier against dust and moisture. The transparent material allows for easy content identification without opening the packaging.
  • Provides a dust and moisture barrier
  • Incorporates a corrosion inhibitor
  • Constructed from low-density polyethylene
  • Transparent material for content visibility
  • Suitable for protecting stored components
  • Blue color for easy identification
  • Eight inch width
  • Four mil thickness
  • Package quantity of 1000 pieces
Catalog No. 50-191-7351 Supplier MSC Supplier No. 39569140
This VCI Poly Tubing offers effective protection for items requiring corrosion inhibition during storage. Made from low-density polyethylene film infused with a corrosion inhibitor, it forms a reliable barrier against dust and moisture. The transparent material allows for easy content identification without opening the packaging.
  • Provides a dust and moisture barrier
  • Incorporates a corrosion inhibitor
  • Constructed from low-density polyethylene
  • Transparent material for content visibility
  • Suitable for protecting stored components
  • Blue color for easy identification
  • Eight inch width
  • Four mil thickness
  • Package quantity of 1000 pieces