Learn More
Tewes Corporation VCI BAG 48.5X42.5X60 2 MIL
SDP
Supplier:  Tewes Corporation 075F00001
Encompass_Preferred
VCI Bags (Vapor Corrosion Inhibitors) are easy to use and provide cost effective proven corrosion protection. Ferrous Yellow bags are a special packaging film containing proprietary Zerust corrosion inhibiting additives developed to prevent the corrosion of ferrous metals. Bags will protect metals for up to 5 years if properly packed and stored. Offer a shelf life of 2 years if stored at 85 degrees F (30 degrees C) or less, out of direct sunlight and in original packaging. 100/ROLL.
Catalog No. NC1841703