Learn More
Tewes Corporation VCI PAPER 48 X 200YARDS 1/ROLL
SDP
Supplier:  Tewes Corporation 300M00006
Encompass_Preferred
VCI Paper is saturated with Zerust VCI on both sides for easy application. Provides effective Multi-metal corrosion protection up to 1 year in the convenience and affordability of a paper product. Use in interleaving applications and to wrap metal materials for shipment, work in progress or storage. Parts are ready for use directly out of the packaging as the paper leaves no residue. Product is fully recyclable, repulpable and made from responsibly managed forestry products.
Catalog No. NC1841704