Learn More
MSC VCI Polybag, 23 in. x 17 in. x 46 in., Blue, 1 ea
Catalog No. 501909865
Encompass
This vapor corrosion inhibitor (VCI) polybag is designed to provide effective protection for metallic items against corrosion during storage and transport. The blue-tinted polyethylene bag creates a protective atmosphere that inhibits rust and oxidation on metal surfaces without the need for additional oils or coatings.
  • Dimensions are 23 in. x 17 in. x 46 in.
  • Provides effective corrosion protection for metallic components
  • Features a blue color for easy identification
  • Constructed from durable polyethylene material
Catalog No. 50-190-9865 Supplier MSC Supplier No. 39476700
This vapor corrosion inhibitor (VCI) polybag is designed to provide effective protection for metallic items against corrosion during storage and transport. The blue-tinted polyethylene bag creates a protective atmosphere that inhibits rust and oxidation on metal surfaces without the need for additional oils or coatings.
  • Dimensions are 23 in. x 17 in. x 46 in.
  • Provides effective corrosion protection for metallic components
  • Features a blue color for easy identification
  • Constructed from durable polyethylene material