Learn More
Uline VCI Shrink Film - 6 Mil, 20 x 100'
Supplier:  Uline S20195
Encompass
Shrink film, polyethylene, 20 x 100' roll, volatile corrosion inhibitor, 6 month outdoor protection, 12 month indoor protection
Catalog No. NC2382846