Learn More
Aviva Systems Biology Corp VCL Antibody - C-terminal region (ARP61099_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61099_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VCL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human VCL. Alternate names: CMD1W, CMH15, MVCLApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-250