Learn More
Medchemexpress LLC HY-15575 100mg , VcMMAE CAS:646502-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15575100MG
Encompass_Preferred
HY-15575 100mg VcMMAE CAS: 646502-53-6 VcMMAE (mc-vc-PAB-MMAE) is a drug-linker conjugate for ADC with potent antitumor activity by using the anti-mitotic agent, monomethyl auristatin E (MMAE, a tubulin inhibitor), linked via the lysosomally cleavable dipeptide, valine-citrulline (vc). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9756