Learn More
Selleck Chemical LLC VcMMAE 5mg
Supplier: Selleck Chemical LLC S66615MG
VcMMAE (mc-vc-PAB-MMAE), a MMAE derivative with valine-citrulline (Vc) linker,is anantibody-drug conjugate (ADC) with potent antitumor activity. MMAE is a synthetic antineoplastic agent and can be efficiently released from VcMMAE in vitro and exert cytotoxic activity. *For Research & Development use only. Product is not intended for drug, household, or human consumption.