Learn More
Aviva Systems Biology Corp VCP Antibody - C-terminal region (ARP59365_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59365_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VCP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human VCP. Alternate names: IBMPFD, MGC131997, MGC148092, MGC8560, TERA, p97, ALS14ApplicationWB, IP, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-675