Learn More
Agilent Technologies VCSM13INTERFERERESISTHELPPHAGE
Catalog No. 50125059
Encompass
VCSM13 Interference-Resistant Helper Phage 1mL VCSM13 helper phage (~1 x 1011 pfu/mL) Interference-resistant helper phage (6 kb single stranded) Derived from M13 K07 mutant Carries kanamycin-resistance gene in IG region
Catalog No. 50-125-059 Supplier Agilent Technologies Supplier No. 200251
VCSM13 Interference-Resistant Helper Phage 1mL VCSM13 helper phage (~1 x 1011 pfu/mL) Interference-resistant helper phage (6 kb single stranded) Derived from M13 K07 mutant Carries kanamycin-resistance gene in IG region