Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Sodium Heparin Additive, 10 mL, Conventional Closure Plastic Tube, 100/Box
Catalog No. 503044730
Encompass_Preferred
Features Sodium Heparin (spray-coated), 150 USP units
Catalog No. 50-304-4730 Supplier Med Vet International Supplier No. 367874
Features Sodium Heparin (spray-coated), 150 USP units