Learn More
Med Vet International BD Vacutainer Barricor, Blood Collection Tube, Plastic, 4.5mL, 13 x 100mm, Lime, 100/Box
Catalog No. 503043228
Encompass_Preferred
BD Barricor tube with mechanical separator and lithium heparin for plasma separation.
Catalog No. 50-304-3228 Supplier Med Vet International Supplier No. 365049
BD Barricor tube with mechanical separator and lithium heparin for plasma separation.