Learn More
Med Vet International BD Vacutainer Barricor, Blood Collection Tube, Plastic, 5.5mL, 13 x 100mm, Lime, 100/Box
Catalog No. 503044713
Encompass_Preferred
BD Barricor tube with mechanical separator and lithium heparin for plasma separation.
Catalog No. 50-304-4713 Supplier Med Vet International Supplier No. 365056
BD Barricor tube with mechanical separator and lithium heparin for plasma separation.