Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Clot Activator Additive, 3 mL, Conventional Closure Plastic Tube, 100/Box
Catalog No. 503044714
Encompass_Preferred
Features Spray dried clot activator (silica,PVP,L-720)
Catalog No. 50-304-4714 Supplier Med Vet International Supplier No. 366668
Features Spray dried clot activator (silica,PVP,L-720)