Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Sodium Fluoride/Potassium Oxalate Additive, 2 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503044731
Encompass_Preferred
Features Sodium Fluoride, 5.0 mg / Potassium Oxalate, 4.0 mg For glucose determination
Catalog No. 50-304-4731 Supplier Med Vet International Supplier No. 367921
Features Sodium Fluoride, 5.0 mg / Potassium Oxalate, 4.0 mg For glucose determination