Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Clot Activator Additive, 4 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503043032
Encompass_Preferred
Features Not recommended for blood bank use Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
Catalog No. 50-304-3032 Supplier Med Vet International Supplier No. 367812
Features Not recommended for blood bank use Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact