Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Sodium Fluoride/Potassium Oxalate Additive, 6 mL, BD Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503043264
Encompass_Preferred
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact For glucose determination
Catalog No. 50-304-3264 Supplier Med Vet International Supplier No. 367925
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact For glucose determination