Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, K2 EDTA Additive, 6 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503043035
Encompass_Preferred
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact K2 EDTA (spray-dried), 10.8mg
Catalog No. 50-304-3035 Supplier Med Vet International Supplier No. 367863
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact K2 EDTA (spray-dried), 10.8mg