Learn More
Med Vet International BD Vacutainer PST™ Venous Blood Collection Tube, Lithium Heparin/Separator Gel Additive, 3 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503043557
Encompass_Preferred
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact Additive: Lithium heparinN (spray coated) and polymer separator gel
Catalog No. 50-304-3557 Supplier Med Vet International Supplier No. 367960
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact Additive: Lithium heparinN (spray coated) and polymer separator gel