Learn More
Med Vet International BD Vacutainer SPC Plus Venous Blood Collection Tube, K2 EDTA Additive, 6 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503043262
Encompass_Preferred
Features BD Medical 368381 Vacutainer® Plus Plastic Blood Collection Tubes for Trace Element Testing Paper Label and Royal Blue Hemogard™ Closure Additive: K?EDTA, 10.8 mg 510(k) Cleared
Catalog No. 50-304-3262 Supplier Med Vet International Supplier No. 368381
Features BD Medical 368381 Vacutainer® Plus Plastic Blood Collection Tubes for Trace Element Testing Paper Label and Royal Blue Hemogard™ Closure Additive: K?EDTA, 10.8 mg 510(k) Cleared