Learn More
Med Vet International BD Vacutainer UltraTouch™ Push Button Blood Collection Set, 21 Gauge, 3/4 Inch Needle Length, Safety Needle, 12 Inch Tubing, Sterile, 50/Box
Catalog No. 503043259
Encompass_Preferred
Features The BD Vacutainer® UltraTouch™ Push Button Blood Collection Set is engineered to minimize patient discomfort during blood collection Pentapoint™ Comfort and RightGauge™ Ultra-Thin Wall technology reduce penetration forces by 32% without compromising on tube fill times or sample quality Singe-handed, in-vein safety activation instantly retracts needle after use
Catalog No. 50-304-3259 Supplier Med Vet International Supplier No. 367365
Features The BD Vacutainer® UltraTouch™ Push Button Blood Collection Set is engineered to minimize patient discomfort during blood collection Pentapoint™ Comfort and RightGauge™ Ultra-Thin Wall technology reduce penetration forces by 32% without compromising on tube fill times or sample quality Singe-handed, in-vein safety activation instantly retracts needle after use