Learn More
Enzo Life Sciences Voltage-dependent anion channel polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210785C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a part (CNDGTEFGGSIYQK) of C-terminal human VDAC (voltage-dependent anion channel) with an added N-terminal cysteine. This sequence is completely conserved between isoforms 1, 2 and 3 of human, mouse and rat VDAC. Species reactivity: Human, Mouse, Rat, Monkey. Applications: IP, WB. Recommended Dilutions: Western Blot (2µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8153