Learn More
Biomatik Corporation Vdac1 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07019
Encompass_Preferred
Vdac1 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to VDAC1. The Vdac1 Recombinant Antibody has been validated for the following applications: ELISA, WB, IHC. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Vdac1. Applications: ELISA, WB, IHC. Target Synonyms: VDAC1. Accession Number: P21796. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7689