Learn More
Aviva Systems Biology Corp VDAC2 Antibody - N-terminal region (ARP35123_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35123_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VDAC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VDAC2. Alternate names: RP11-375G3.1, FLJ23841, PORApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-55648