Learn More
ABclonal Technology VDAC2 Rabbit mAb
SDP
Catalog No. 5000006784
Encompass_Preferred
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist killer 1 protein activity Pseudogenes have been identified on chromosomes 1 2 12 and 21 and alternative splicing results in multiple transcript variants
Catalog No. 50-000-06784 Supplier ABclonal Technology Supplier No. A21260100UL
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist killer 1 protein activity Pseudogenes have been identified on chromosomes 1 2 12 and 21 and alternative splicing results in multiple transcript variants